Synonyms:
4-(4-Maleimidophenyl)butyric Acid N-Succinimidyl EsterSMPB
S0861-10MG
TCS0861-10MGEA
137
USD
InStock
TCS0861-10MG
N-Succinimidyl 4-(4-Maleimidophenyl)butyrate (SMPB) ≥97.0% (by HPLC)
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)
CAS Number: 79886-55-8
MDL Number: MFCD00054961
Molecular Formula: C18H16N2O6
Molecular Weight: 356.33
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 131
Storage Temperature: 0°C
Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: 4-(4-Maleimidophenyl)butyric Acid N-Succinimidyl Ester, SMPB