Synonyms:
Sodium peroxometaborate tetrahydrate
S0887-25G
S0887-500G
TCS0887-500GEA
45.62
USD
InStock
TCS0887-500G
TCS0887-25G
Sodium perborate tetrahydrate ≥97.0% (by titrimetric analysis) for Biochemical Research
Sodium perborate tetrahydrate
[for Biochemical Research]
CAS Number: 10486-00-7
MDL Number: MFCD00149231
Molecular Formula: BNaO3
Molecular Weight: 81.80
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 60