Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfamide ≥96.0% (by total nitrogen basis)

Supplier: TCI America
Synonyms: Sulphuric diamideSulfuric diamide

S0890-25G S0890-250G
TCS0890-25GEA 30.02 USD InStock
TCS0890-25G TCS0890-250G
Sulfamide ≥96.0% (by total nitrogen basis)
Sulfamide
CAS Number: 7803-58-9
MDL Number: MFCD00011606
Molecular Formula: H4N2O2S
Molecular Weight: 96.10
Purity/Analysis Method: 96.0% (N)
Form: Crystal
Color: White
Melting point (°C): 91


Formula: H₄N₂O₂S
MW: 96.11 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00011606
CAS Number: 7803-58-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/42/78/28994278.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty