Synonyms:
Silver Triflimide
S0898-1G
S0898-5G
TCS0898-1GEA
138.27
USD
InStock
TCS0898-1G
TCS0898-5G
Silver bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Silver bis(trifluoromethanesulfonyl)imide
CAS Number: 189114-61-2
MDL Number: MFCD08457644
Molecular Formula: C2AgF6NO4S2
Molecular Weight: 388.00
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 249
Storage Temperature: 0°C
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, C-H Bond Activation Reaction, Additives ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Silver Triflimide