Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α',α'-Tetrabromo-o-xylene ≥96.0% (by GC)

Supplier: TCI America
Synonyms: 1,2-Bis(dibromomethyl)benzene

T0052-25G
TCT0052-25GEA 61.87 USD InStock
TCT0052-25G
α,α,α',α'-Tetrabromo-o-xylene ≥96.0% (by GC)
α,α,α',α'-Tetrabromo-o-xylene
CAS Number: 13209-15-9
MDL Number: MFCD00000131
Molecular Formula: C8H6Br4
Molecular Weight: 421.75
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 115

Other Product Names / Synonyms: 1,2-Bis(dibromomethyl)benzene, o-Xylylene Tetrabromide


Formula: C₈H₆Br₄
MW: 421.75 g/mol
Melting Pt: 115 °C
MDL Number: MFCD00000131
CAS Number: 13209-15-9
UN: 3143
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/93/28994393.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty