Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrapropylammonium hydroxide 10% in water

Supplier: TCI America
Synonyms: IPC-TPA-OH

T0171-100ML T0171-25ML
TCT0171-100MLEA 98.46 USD InStock
TCT0171-100ML TCT0171-25ML
Tetrapropylammonium hydroxide 10% in water
Tetrapropylammonium hydroxide
CAS Number: 4499-86-9
MDL Number: MFCD00009360
Molecular Formula: C12H29NO
Molecular Weight: 203.37
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.00


Formula: C₁₂H₂₉NO
MW: 203.37 g/mol
Density: 1.00 (20 °C)
MDL Number: MFCD00009360
CAS Number: 4499-86-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/91/28994491.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty