T0185-5G
T0185-25G
TCT0185-5GEA
24.36
USD
InStock
TCT0185-5G
TCT0185-25G
Thiazole ≥98.0% (by GC)
Thiazole
CAS Number: 288-47-1
MDL Number: MFCD00005315
Molecular Formula: C3H3NS
Molecular Weight: 85.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 118
Melting point (°C): -33
Flash Point (°C): 26
Specific Gravity (20/20): 1.20