Synonyms:
1,3-Trimethylene Oxide
, Trimethylene oxide
T0473-10ML
TCT0473-10MLEA
306
USD
InStock
TCT0473-10ML
Oxetane ≥98.0% (by GC)
Oxetane
CAS Number: 503-30-0
MDL Number: MFCD00005167
Molecular Formula: C3H6O
Molecular Weight: 58.08
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 50
Flash Point (°C): -28
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1,3-Epoxypropane, Oxetane