T0792-25G
TCT0792-25GEA
93.76
USD
InStock
TCT0792-25G
Trichloroacetic anhydride ≥97.0% (by titrimetric analysis)
Trichloroacetic anhydride
CAS Number: 4124-31-6
MDL Number: MFCD00000793
Molecular Formula: C4Cl6O3
Molecular Weight: 308.74
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 116
Specific Gravity (20/20): 1.69
Other Product Names / Synonyms: Perchloroacetic Anhydride