Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotrimethylgermane ≥98.0% (by GC)

Supplier: TCI America
Synonyms: ChlorotrimethylgermaneTrimethylgermanium chloride

T1161-1G
TCT1161-1GEA 172.55 USD InStock
TCT1161-1G
Chlorotrimethylgermane ≥98.0% (by GC)
Chlorotrimethylgermane
CAS Number: 1529-47-1
MDL Number: MFCD00000462
Molecular Formula: C3H9ClGe
Molecular Weight: 153.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 102
Flash Point (°C): 1
Specific Gravity (20/20): 1.25

Other Product Names / Synonyms: Trimethylgermanium Chloride


Formula: C₃H₉ClGe
Boiling Pt: 102 °C
Density: 1.25 (20 °C)
Flash Pt: 1 °C
MDL Number: MFCD00000462
CAS Number: 1529-47-1
UN: 2924
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/52/17/28995217.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty