Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α,2,3,5,6-Heptafluorotoluene ≥97.0% (by GC)

Supplier: TCI America
Synonyms: 1,2,4,5-Tetrafluoro-3-(trifluoromethyl)benzene 2,3,5,6-Tetrafluorobenzotrifluoride

T1854-5G
TCT1854-5GEA 73.42 USD InStock
TCT1854-5G
α,α,α,2,3,5,6-Heptafluorotoluene ≥97.0% (by GC)
α,α,α,2,3,5,6-Heptafluorotoluene
CAS Number: 651-80-9
MDL Number: MFCD00012385
Molecular Formula: C7HF7
Molecular Weight: 218.07
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 32
Specific Gravity (20/20): 1.61
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,2,3,5,6-Heptafluorotoluene


Formula: C₇HF₇
MW: 218.07 g/mol
Density: 1.61 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00012385
CAS Number: 651-80-9
UN: 1993
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/58/24/28995824.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty