Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethylamine ∼13% ∼2 mol/L in THF

Supplier: TCI America

T2704-500ML
TCT2704-500MLEA 127.58 USD InStock
TCT2704-500ML
Trimethylamine ∼13% ∼2 mol/L in THF
Trimethylamine
CAS Number: 75-50-3
MDL Number: MFCD00008327
Molecular Formula: C3H9N
Molecular Weight: 59.11
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0-10°C


Formula: C₃H₉N
MW: 59.11 g/mol
MDL Number: MFCD00008327
CAS Number: 75-50-3
UN: 1297
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/63/97/28996397.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty