Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Triphenylphosphine)gold(I) chloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Chlorotriphenylphosphine gold(I)Chloro(triphenylphosphine)gold(I)

T2994-1G T2994-200MG
TCT2994-1GEA 184.55 USD InStock
TCT2994-1G TCT2994-200MG
(Triphenylphosphine)gold(I) chloride ≥95.0% (by titrimetric analysis)
(Triphenylphosphine)gold (I) chloride
CAS Number: 14243-64-2
MDL Number: MFCD00009588
Molecular Formula: C18H15AuClP
Molecular Weight: 494.71
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 247

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Metal Catalysts ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Chloro(triphenylphosphine)gold(I)


Formula: C₁₈H₁₅AuClP
MW: 494.71 g/mol
Melting Pt: 247 °C
MDL Number: MFCD00009588
CAS Number: 14243-64-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/66/14/28996614.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty