T3011-1G
T3011-5G
TCT3011-1GEA
103.48
USD
InStock
TCT3011-1G
TCT3011-5G
(Pentafluoroethyl)trimethylsilane ≥98.0% (by GC)
(Pentafluoroethyl)trimethylsilane
CAS Number: 124898-13-1
MDL Number: MFCD04116436
Molecular Formula: C5H9F5Si
Molecular Weight: 192.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Specific Gravity (20/20): 1.11
Product Categories: Synthetic Organic Chemistry, Other Fluorinated Group Introducing Agents
Other Product Names / Synonyms: Trimethyl(perfluoroethyl)silane