T3178-200MG
TCT3178-200MGEA
26.38
USD
InStock
TCT3178-200MG
2,4,6-Tris(4-bromophenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tris(4-bromophenyl)-1,3,5-triazine
CAS Number: 30363-03-2
MDL Number: MFCD04116312
Molecular Formula: C21H12Br3N3
Molecular Weight: 546.06
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 361