Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanillin in ethanol (contains H₂SO₄)

Supplier: TCI America
Synonyms: 4-Hydroxy-3-methoxybenzoic acid

V0080-500ML
TCV0080-500MLEA 153.51 USD InStock
TCV0080-500ML
Vanillin in ethanol (contains H₂SO₄)
Vanillin
[for TLC Stain]
CAS Number: 121-33-5
MDL Number: MFCD00006942
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 153
Storage Temperature: 0-10°C


Formula: C₈H₈O₄
MW: 152.15 g/mol
Flash Pt: 153 °C
MDL Number: MFCD00002551
CAS Number: 121-33-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/73/11/28997311.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty