Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc chloride ∼7% ∼0.5 mol/L in THF

Supplier: TCI America
Synonyms: Zinc (II) chlorideButter of zinc

Z0019-100ML
TCZ0019-100MLEA 111.65 USD InStock
TCZ0019-100ML
Zinc chloride ∼7% ∼0.5 mol/L in THF
Zinc chloride
CAS Number: 7646-85-7
MDL Number: MFCD00011295
Molecular Formula: Cl2Zn
Molecular Weight: 136.28
Form: Clear Liquid
Color: Colorless


Formula: ZnCl₂
MW: 136.3 g/mol
MDL Number: MFCD00011295
CAS Number: 7646-85-7
UN: 2331
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/38/28997438.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty