Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc(II) Bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Zinc bis(trifluoromethylsulfonyl)imideZinc bis(trifluoromethylsulphonyl)imideBis(trifluoromethanesulfonyl)imide Zinc(II) SaltBis(trifluoromethanesulphonyl)imide Zinc(II) SaltZinc(II) Triflimide

Z0027-1G Z0027-5G
TCZ0027-5GEA 313.33 USD InStock
TCZ0027-5G TCZ0027-1G
Zinc(II) Bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Zinc(II) Bis(trifluoromethanesulfonyl)imide
CAS Number: 168106-25-0
MDL Number: MFCD16621474
Molecular Formula: C4F12N2O8S4Zn
Molecular Weight: 625.65
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 147
Flash Point (°C): 110

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Bis(trifluoromethanesulfonyl)imide Zinc(II) Salt, Zinc(II) Triflimide


Formula: C₄F₁₂N₂O₈S₄Zn
MW: 625.65 g/mol
Melting Pt: 147 °C
Flash Pt: 110 °C
MDL Number: MFCD16621474
CAS Number: 168106-25-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/45/28997445.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty