Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Jomac® Kevlar Terrycloth Gloves, Loop Out, Wells Lamont

Supplier: Wells Lamont
636KCL
80091-482EA 129.79 USD InStock
80091-482
Jomac® Kevlar Terrycloth Gloves, Loop Out, Wells Lamont
Gloves
Jomac Kevlar Terry Cloth Gloves resist heat up to 700 °F.

The ambidextrous gloves feature gauntlet cuffs (5"), cotton lining for comfort, and a Clute pattern.

The Kevlar outer shell provides cut and heat resistance.

Certifications: ANSI Level 4.

Ordering information: Gloves are sold by the dozen pairs.

Caution: Caution Information: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty