Would you like to log in again?
Glass Fiber Filters for Air Monitoring, Advantec MFS
Supplier: Advantec
GB100R102MM GB100R142MM GC508X10IN GB100R47MM GB100R127MM GB100R125MM GB100R25MM GB100R293MM GB100R8X10IN GB100R55MM GB100R124MM GB100R150MM GB100R90MM
102831-102PK 784.87 USD InStock
102831-102 101103-090 102831-096 102831-094 102831-100 102831-098 101103-406 101103-074 101103-094 101103-056 101103-472 101103-076 101103-088
Glass Fiber Filters for Air Monitoring, Advantec MFS
Filters Glass Fiber Filters
Advantec glass fiber filters are made of pure borosilicate glass microfibers.
Grade GB100R filters with very low trace metal content of As, Pb, Cd used in high and low volume air samplers for aerosols, airborne dust and metal contaminants. Gas collection efficiency (DOP) of 99.99%. Other applications include filtering DNA/RNA and protein precipitates.
Grade GC50 thin filter with high collection efficiency (DOP) of 99.99% suitable as prefilter for membrane. Low absorption filter for scintillation counting.
Grade GA55 for air pollution monitoring and clarifying filtration.
Highly resistant to chemical attack and biologically inert. Binder-free.
Ordering information: Other diameters available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty