Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Promega
V8051
PAV8051EA 198.83 USD InStock
PAV8051
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Polyclonal secondary antibody raised in donkey, immunoaffinity-purified using immobilized antigens and conjugated to horseradish peroxidase (HRP) enzyme.

  • Secondary Antibody That Reacts with Goat and Sheep IgG
  • Extensively used and published in numerous research articles
  • Antibody is ready-to-use

Polyclonal secondary antibody raised in donkey, immunoaffinity-purified using immobilized antigens and conjugated to horseradish peroxidase (HRP) enzyme. The Donkey Anti-Goat IgG, HRP Conjugate is a secondary antibody developed in donkeys against goat IgG. Donkey Anti-Goat IgG, HRP Conjugate shows reactivity to goat and sheep IgG but minimal cross-reactivity to rabbit and mouse IgG. For Western blot applications with chromogenic detection use at a starting dilution of 1:10,000. As with all antibodies, in certain applications some species-dependent antigen-dependent cross-reactivity may be observed. The product (unless otherwise noted) is supplied as a 1mg/ml solution.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty