Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Promega
W4011
PAW4011EA 159.39 USD InStock
PAW4011
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Polyclonal secondary antibody raised in goat, immunoaffinity-purified using immobilized antigens and conjugated to horseradish peroxidase (HRP) enzyme. The rabbit antibodies react with all IgG subclasses.

  • Secondary Antibody That Binds to Both Heavy and Light Chains for All Rabbit IgG Subclasses
  • Extensively used and published in numerous research articles
  • Antibody is ready-to-use

Polyclonal secondary antibodies raised in goat, immunoaffinity-purified using immobilized antigens and conjugated to horseradish peroxidase (HRP) enzyme. The Anti-Rabbit IgG (H+L), HRP Conjugate, antibody binds to both heavy and light chains for all IgG subclasses. As with all antibodies, in certain applications some species-dependent antigen-dependent cross-reactivity may be observed. The products (unless otherwise noted) are supplied as 1mg/ml solutions. A starting working dilution of 1:2,500 is suggested for most Western blot, dot blot and ELISA applications. The optimum concentration of secondary antibody depends on the application and will need to be empirically determined.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty