Would you like to log in again?
Apex Superior Adhesive Slides, Leica
Supplier: Leica
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Surgipath®
3800083 3800085 3800084 3800081 3800082 3800080 3800075 3800086
75806-640CS 763.65 USD InStock
75806-640 10015-146 75806-642 10015-148 10015-152 75806-644 10015-150 10015-162
Apex Superior Adhesive Slides, Leica
Slides Adhesive Slides
The Apex Adhesive Slides portfolio consists of microscope slides made of white glass and coated with a proprietary adhesive coating for a positive charge.
  • White glass construction
  • Coated for a positive charge
  • Hydrophilic for homogeneous covering by aqueous stains and reagents
  • Consistent, full slide staining
The charge promotes tissue adhesion and allows the slide to be more hydrophilic. This hydrophilicity provides a better base for homogeneous covering by aqueous stains and reagents, leading to consistent, full slide staining. All slides in the Apex portfolio are excellent for demanding procedures such as standard IHC and HIER techniques.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty