Would you like to log in again?
Premier Cover Glass, Leica
Supplier: Leica
Surgipath®
3800150ACS 3800145ACS 3800160ACS 3800140ACS
10015-168CS 85.85 USD InStock
10015-168 10015-166 10015-170 75810-254
Premier Cover Glass, Leica
Cover Glasses
Premier Cover Glass has been specially designed to work with automated coverslip machines.
  • Work with automated coverslip machines
  • Prevent sticking
  • Available in multiple size options
Extra processes to prevent sticking are employed during manufacturing procedures. The cover glass is available in several sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty