Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 95.0-98.0% ACS

Supplier: BeanTown Chemical

121240-1KG
BT121240-1KGEA 64.79 USD InStock
BT121240-1KG
Sulfuric acid 95.0-98.0% ACS
Sulfuric acid
  • RTECS: WS5600000
  • Liquid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 290 °C
Melting Pt: 3 °C
Density: 1.84 g/mL
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Assay 95.0-98.0%
Color (APHA) ≤10
Residue after ignition ≤5ppm
Nitrate (NO3) ≤0.5ppm
Chloride (Cl) ≤0.2ppm
Arsenic (As) ≤0.01ppm
Ammonium (NH4) ≤2ppm
Substances reducing KMnO4 Passes test
Heavy metals (as Pb) ≤1ppm
Iron (Fe) ≤0.2ppm
Mercury (Hg) ≤5ppb

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty