Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-chloropropane ≥99%

Supplier: BeanTown Chemical

121325-100G 121325-250G 121325-1KG
BT121325-250GEA 25.96 USD InStock
BT121325-250G BT121325-100G BT121325-1KG
1-Bromo-3-chloropropane ≥99%
1-Bromo-3-chloropropane
  • RTECS: TX4113000
  • Refractive index: 1.486
  • Liquid


Formula: BrH₂CCH₂CH₂Cl
MW: 157.44 g/mol
Boiling Pt: 144…145 °C
Density: 1.592 g/mL
Flash Pt: 45 °C (113 °F)
MDL Number: MFCD00000998
CAS Number: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty