Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Cyanostyrene ≥97% stabilized

Supplier: BeanTown Chemical
Synonyms: 4-Ethenylbenzonitrile

122645-5G 122645-1G 122645-25G
BT122645-5GEA 229.54 USD InStock
BT122645-5G BT122645-25G BT122645-1G
4-Cyanostyrene ≥97% stabilized
4-Cyanostyrene
  • Liquid


Formula: C₉H₇N
MW: 129.162 g/mol
MDL Number: MFCD00080445
CAS Number: 3435-51-6
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty