Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroacetic acid ≥99% ACS

Supplier: BeanTown Chemical

122765-500G 122765-100G
BT122765-500GEA 113.76 USD InStock
BT122765-500G BT122765-100G
Chloroacetic acid ≥99% ACS
Chloroacetic acid
  • RTECS: AF8575000
  • Hygroscopic
  • Crystalline


Formula: ClCH₂COOH
MW: 94.5 g/mol
Boiling Pt: 189 °C
Melting Pt: 60…63 °C
Flash Pt: 126 °C (259 °F)
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/30/2302030.jpg
Order Now
Specification Test Results
Assay 99%
Loss on drying ≤2.7%
Titrable acid ≤0.020meq/g
Arsenic (As) ≤0.015%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty