Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-methylpropane ≥98%

Supplier: BeanTown Chemical
Synonyms: Isobutyl bromide

122785-250G 122785-1KG
BT122785-1KGEA 141.24 USD InStock
BT122785-1KG BT122785-250G
1-Bromo-2-methylpropane ≥98%
1-Bromo-2-methylpropane
  • RTECS: TX4140000
  • Refractive index: 1.435
  • Liquid


Formula: C₄H₉Br
MW: 137.02 g/mol
Boiling Pt: 90…92 °C
Density: 1.26 g/mL
Flash Pt: 18 °C (64 °F)
MDL Number: MFCD00000217
CAS Number: 78-77-3
EINECS: 201-141-2
UN: 2342
ADR: 3,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty