Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butylamine ≥98%

Supplier: BeanTown Chemical
Synonyms: 2-Aminoisobutane

122865-2.5L 122865-500ML 122865-10L
BT122865-500MLEA 29.18 USD InStock
BT122865-500ML BT122865-10L BT122865-2.5L
tert-Butylamine ≥98%
tert-Butylamine
  • RTECS: EO3330000
  • Refractive index: 1.377
  • Air Sensitive
  • Liquid


Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 46 °C
Melting Pt: –67 °C
Density: 0.696 g/mL
Flash Pt: –38 °C (–36 °F)
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2733
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty