Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraammineplatinum(II) nitrate

Supplier: BeanTown Chemical

123655-1G 123655-5G
BT123655-5GEA 727.09 USD InStock
BT123655-5G BT123655-1G
Tetraammineplatinum(II) nitrate
Tetraammineplatinum(II) nitrate
  • Powder/Chunks


Formula: H₁₂N₆O₆Pt
Melting Pt: 262 °C (dec.)
MDL Number: MFCD00011624
CAS Number: 20634-12-2
EINECS: 243-929-9
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty