Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crotonic acid

Supplier: BeanTown Chemical
Synonyms: 2-Butenoic acidtrans-2-Butenoic acidtrans-Crotonic acid

123965-500G
BT123965-500GEA 49.44 USD InStock
BT123965-500G
Crotonic acid
Crotonic acid
  • RTECS: GQ2900000
  • Flakes


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 180…181 °C
Melting Pt: 70…72 °C
Density: 1.027 g/mL
Flash Pt: 88 °C (190 °F)
MDL Number: MFCD00002701
CAS Number: 107-93-7
EINECS: 203-533-9
UN: 2823
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty