Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) thiocyanate

Supplier: BeanTown Chemical
Synonyms: Mercuric thiocyanateMercury (II) rhodanideMercury dithiocyanate

124015-250G 124015-50G
BT124015-250GEA 259.66 USD InStock
BT124015-250G BT124015-50G
Mercury(II) thiocyanate
Mercury (II) thiocyanate
  • RTECS: XL1550000
  • Light Sensitive, Moisture Sensitive
  • Powder


Formula: C₂HgN₂S₂
MW: 316.76 g/mol
Melting Pt: 165 °C (dec.)
Density: 3.71 g/mL
Flash Pt: 120 °C (248 °F)
MDL Number: MFCD00011039
CAS Number: 592-85-8
EINECS: 209-773-0
UN: 1646
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty