Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Arsenic ≥99.99% (trace metals basis), powder -70 mesh

Supplier: BeanTown Chemical

124535-10G 124535-50G
BT124535-10GEA 162.58 USD InStock
BT124535-10G BT124535-50G
Arsenic ≥99.99% (trace metals basis), powder -70 mesh
Arsenic
  • RTECS: CG0525000
  • Powder


Formula: As
MW: 74.92 g/mol
Boiling Pt: 613 °C (subl.)
Melting Pt: 817 °C
Density: 5.727 g/mL
MDL Number: MFCD00085309
CAS Number: 7440-38-2
EINECS: 213-148-6
UN: 1558
ADR: 6.1,II
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty