Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl chloroformate ≥95% stabilized

Supplier: BeanTown Chemical
Synonyms: CBZ-Cl (Z-Cl)

124690-1KG 124690-250G 124690-50G
BT124690-1KGEA 657.5 USD InStock
BT124690-1KG BT124690-50G BT124690-250G
Benzyl chloroformate ≥95% stabilized
Benzyl chloroformate
  • Refractive index: 1.519
  • Moisture Sensitive
  • Liquid


Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 103 °C/20 mmHg
Density: 1.195 g/mL
Flash Pt: 92 °C (198 °F)
MDL Number: MFCD00000640
CAS Number: 501-53-1
EINECS: 207-925-0
UN: 1739
ADR: 8,I
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty