Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) fluoride ≥99%

Supplier: BeanTown Chemical
Synonyms: Manganese(II) difluorideManganese difluoride

125245-50G 125245-10G
BT125245-50GEA 99.2 USD InStock
BT125245-50G BT125245-10G
Manganese(II) fluoride ≥99%
Manganese (II) fluoride
  • RTECS: OP0875000
  • Hygroscopic
  • Crystalline/Powder


Formula: F₂Mn
MW: 92.93 g/mol
Melting Pt: 856 °C
Density: 3.98 g/mL
MDL Number: MFCD00016222
CAS Number: 7782-64-1
EINECS: 231-960-0
UN: 3288
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty