Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hexafluorotitanate ≥97%

Supplier: BeanTown Chemical
Synonyms: Potassium hexafluorotitanate(IV)

125275-500G 125275-100G 125275-2.5KG
BT125275-500GEA 55.47 USD InStock
BT125275-500G BT125275-2.5KG BT125275-100G
Potassium hexafluorotitanate ≥97%
Potassium hexafluorotitanate
  • RTECS: TT1575000
  • Powder


Formula: F₆K₂Ti
MW: 240.07 g/mol
Melting Pt: 780 °C
MDL Number: MFCD00011380
CAS Number: 16919-27-0
EINECS: 240-969-9
UN: 3288
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty