Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium(II) nitrate (1% Pd) in nitric acid 10% Matrix Modifier Solution

Supplier: BeanTown Chemical

125290-100ML
BT125290-100MLEA 411.33 USD InStock
BT125290-100ML
Palladium(II) nitrate (1% Pd) in nitric acid 10% Matrix Modifier Solution
Palladium (II) nitrate
  • Liquid, 1% Pd in 10% HNO3


Formula: N₂O₆Pd
MW: 230.43 g/mol
Density: 1.11 g/mL
MDL Number: MFCD00011169
CAS Number: 10102-05-3
EINECS: 231-714-2
UN: 3264
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty