Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethynyltributyltin ≥95%

Supplier: BeanTown Chemical
Synonyms: Ethynyltri-n-butylstannaneTributylethynylstannane

126385-5G 126385-1G
BT126385-5GEA 256.53 USD InStock
BT126385-5G BT126385-1G
Ethynyltributyltin ≥95%
Ethynyltributyltin
  • Refractive index: 1.476
  • Moisture Sensitive
  • Liquid


Formula: C₁₄H₂₈Sn
MW: 315.09
Boiling Pt: 70 °C/0.2 mmHg
Density: 1.089 g/mL
Flash Pt: 74 °C (165 °F)
MDL Number: MFCD00009420
CAS Number: 994-89-8
UN: 2788
ADR: 6.1,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty