Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylimidazole ≥97%

Supplier: BeanTown Chemical
Synonyms: 2-Methyl-1H-imidazole

126915-100G 126915-500G 126915-2.5KG
BT126915-500GEA 64.81 USD InStock
BT126915-500G BT126915-2.5KG BT126915-100G
2-Methylimidazole ≥97%
2-Methylimidazole
  • RTECS: NI7175000
  • Crystalline/Powder


Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 267…268 °C
Melting Pt: 142…143 °C
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 1759
ADR: 8,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty