Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tellurium(IV) ethoxide ≥85%, Technical Grade

Supplier: BeanTown Chemical

127010-1G
BT127010-1GEA 35.09 USD InStock
BT127010-1G
Tellurium(IV) ethoxide ≥85%, Technical Grade
Tellurium(IV) ethoxide
  • Light Sensitive, Moisture Sensitive
  • Liquid


Formula: Te(OC₂H₅)₄
MW: 307.84 g/mol
Boiling Pt: 88…90 °C/2 mmHg
Melting Pt: >100 °C (dec.)
Density: 1.52 g/mL
Flash Pt: 27 °C (81 °F)
MDL Number: MFCD00270937
CAS Number: 2017-01-8
ADR: 3,III
Order Now
Specification Test Results
Assay 85%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty