Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylhydrazinium chloride ≥99%

Supplier: BeanTown Chemical
Synonyms: Phenylhydrazinium chloridePhenylhydrazine hydrochloride

127140-100G
BT127140-100GEA 88.14 USD InStock
BT127140-100G
Phenylhydrazinium chloride ≥99%
Phenylhydrazinium chloride
  • RTECS: MV9000000
  • Air Sensitive, Hygroscopic, Light Sensitive
  • Solid


Formula: C₆H₉ClN₂
MW: 144.6 g/mol
Melting Pt: 250…254 °C (dec.)
MDL Number: MFCD00012924
CAS Number: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/26/2297426.jpg
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty