Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) sulfide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Lead sulphideLead sulfide

127945-50G 127945-10G
BT127945-10GEA 68.55 USD InStock
BT127945-10G BT127945-50G
Lead(II) sulfide ≥99.9% (trace metals basis)
Lead (II) sulphide
  • RTECS: OG4550000
  • -200 Mesh Powder


Formula: ̍PbS
MW: 239.27 g/mol
Density: 7.7 g/mL
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty