Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dihydrogen dinitrosulfatoplatinate(II) (4 - 6% Pt) (w/w) in water

Supplier: BeanTown Chemical

128210-C500MG 128210-C10G 128210-C2G
BT128210-C10GEA 1525.87 USD InStock
BT128210-C10G BT128210-C2G BT128210-C500
Dihydrogen dinitrosulfatoplatinate(II) (4 - 6% Pt) (w/w) in water
Dihydrogen dinitrosulfatoplatinate(II)
  • Liquid


Formula: H₂Pt(NO₂)₂SO₄
MW: 384.17 g/mol (anhy)
MDL Number: MFCD00064668
CAS Number: 12033-81-7
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty