Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyltrichlorosilane ≥95%

Supplier: BeanTown Chemical

128550-25G 128550-5G
BT128550-5GEA 40.3 USD InStock
BT128550-5G BT128550-25G
Allyltrichlorosilane ≥95%
Allyltrichlorosilane
  • RTECS: VV1530000
  • Refractive index: 1.443
  • Moisture Sensitive
  • Liquid


Formula: H₂CCHCH₂SiCl₃
MW: 175.52 g/mol
Boiling Pt: 116 °C/750 mmHg
Density: 1.211 g/mL
Flash Pt: 18 °C (64 °F)
MDL Number: MFCD00000486
CAS Number: 107-37-9
EINECS: 203-485-9
UN: 1724
ADR: 8,II
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty