Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexane ≥99%

Supplier: BeanTown Chemical
Synonyms: Hexahydro-BenzeneHexamethylene

128715-1L 128715-4L
BT128715-1LEA 56.92 USD InStock
BT128715-1L BT128715-4L
Cyclohexane ≥99%
Cyclohexane
  • RTECS: GU6300000
  • Refractive index: 1.426
  • Liquid


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 80.7 °C
Melting Pt: 4…7 °C
Density: 0.779 g/mL
Flash Pt: –18 °C (–1 °F)
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty