Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.7% environmental

Supplier: BeanTown Chemical
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

129255-1L 129255-4L 129255-4X1L
BT129255-1LEA 69.36 USD InStock
BT129255-1L BT129255-4L BT129255-4X1L
Acetonitrile ≥99.7% environmental
Acetonitrile
  • RTECS: AL7700000
  • Refractive index: 1.344
  • Liquid


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81…82 °C
Melting Pt: –48 °C
Density: 0.786 g/mL
Flash Pt: 2 °C (36 °F)
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Assay 99.7% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty