Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorosulfuric acid ≥99%

Supplier: BeanTown Chemical
Synonyms: Chlorosulphonic acidChlorosulfonic acid

129270-250G 129270-1KG
BT129270-1KGEA 302.21 USD InStock
BT129270-1KG BT129270-250G
Chlorosulfuric acid ≥99%
Chlorosulphuric acid
  • RTECS: FX5730000
  • Refractive index: 1.433
  • Moisture Sensitive
  • Liquid


Formula: HClO₃S
MW: 116.52 g/mol
Boiling Pt: 151…152 °C/755 mmHg
Density: 1.753 g/mL
MDL Number: MFCD00011523
CAS Number: 7790-94-5
EINECS: 232-234-6
UN: 1754
ADR: 8,I
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty