Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium periodate 99.8-100.3% ACS

Supplier: BeanTown Chemical
Synonyms: Potassium metaperiodatePotassium tetroxoiodate (VII)

129840-100G 129840-25G
BT129840-100GEA 100.23 USD InStock
BT129840-100G BT129840-25G
Potassium periodate 99.8-100.3% ACS
Potassium periodate
  • Hygroscopic
  • Powder


Formula: KlO₄
MW: 230 g/mol
Melting Pt: 582 °C
Density: 3.618 g/mL
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Assay 99.8-100.3%
Loss on drying ≤0.10%
Other halogens (as Cl) ≤0.01%
Manganese (Mn) ≤1ppm
Sulfate (SO4) ≤0.01%
Aqueous solubility, in 100ml at 20°C 0.42g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty