Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium tert-butoxide ≥90%, Technical Grade

Supplier: BeanTown Chemical

131060-50G 131060-250G 131060-10G
BT131060-50GEA 118.4 USD InStock
BT131060-50G BT131060-10G BT131060-250G
Magnesium tert-butoxide ≥90%, Technical Grade
Magnesium tert-butoxide
  • Air Sensitive, Moisture Sensitive
  • Powder


Formula: C₈H₁₈MgO₂
MW: 170.535 g/mol
MDL Number: MFCD03788761
CAS Number: 32149-57-8
EINECS: 250-931-3
ADR: 8 (4.1),II
Order Now
Specification Test Results
Assay 90%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty